About this item
Removable deep freeze labels for vials, tubes, and freezer boxes. These thermal transfer freezer labels have a glove-friendly adhesive that ensures the labels can be easily removed without leaving any adhesive residue.
- Temperature range: –80 to 100 ℃ (–112 to 212 ℉)
- Ideal for long-term storage in deep-freeze conditions (–80 ℃)
- Removable adhesive ensures the labels can be easily removed without leaving any adhesive residue
- Printable with all major thermal-transfer printer brands
- Resistant to gamma irradiation (tested up to 50 KGy)
Deep freeze, removable barcode labels for vials, tubes, as well as polypropylene and cardboard boxes. The freezer labels have a strong, glove-friendly adhesive that allows them to adhere firmly to containers in (–20 ℃, –40 ℃, –80 ℃) freezers and dry ice while still being easily removable at room temperature without leaving behind any residue.
Waterproof and tearproof, these removable deep freeze labels can also withstand temperatures ranging up to 100 ℃, making them suitable for general lab use such as the identification of beakers, flasks, boxes, plastic and glass bottles that are stored in fridges and freezers.
66 Options Available Below
Product Details & Documents
Removable deep freeze labels for vials, tubes, and freezer boxes. These thermal transfer freezer labels have a glove-friendly adhesive that ensures the labels can be easily removed without leaving any adhesive residue.
- Temperature range: –80 to 100 ℃ (–112 to 212 ℉)
- Ideal for long-term storage in deep-freeze conditions (–80 ℃)
- Removable adhesive ensures the labels can be easily removed without leaving any adhesive residue
- Printable with all major thermal-transfer printer brands
- Resistant to gamma irradiation (tested up to 50 KGy)
Deep freeze, removable barcode labels for vials, tubes, as well as polypropylene and cardboard boxes. The freezer labels have a strong, glove-friendly adhesive that allows them to adhere firmly to containers in (–20 ℃, –40 ℃, –80 ℃) freezers and dry ice while still being easily removable at room temperature without leaving behind any residue.
Waterproof and tearproof, these removable deep freeze labels can also withstand temperatures ranging up to 100 ℃, making them suitable for general lab use such as the identification of beakers, flasks, boxes, plastic and glass bottles that are stored in fridges and freezers.
Product Family Options
Circle removable deep freeze labels for thermal transfer printers
Product Information
- ColorCore sizeDimensionsPackagingAvailabilityPrice
- 76273-818Deep freeze labelDeep freeze labelWhite1"19.1 mm (0.75")2000 labels/roll (1 label across)Log in for availabilityRoll of 1$221.88Supplier Number: 807656016511
- 76273-820Deep freeze labelDeep freeze labelWhite3"19.1 mm (0.75")2000 labels/roll (1 label across)Log in for availabilityRoll of 1$221.88Supplier Number: 807656016528
- 76273-822Deep freeze labelDeep freeze labelWhite1"25.4 mm (1")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$99.36Supplier Number: 807656016535
- 76273-824Deep freeze labelDeep freeze labelWhite3"25.4 mm (1")5000 labels/roll (1 label across)Log in for availabilityRoll of 1$496.09Supplier Number: 807656016559
Rectangle and circle removable deep freeze labels for thermal transfer printers
Product Information
- ColorCore sizeDimensionsPackagingAvailabilityPrice
- 76273-806Deep freeze labelDeep freeze labelWhite1"23.9×12.7+11.1 mm (0.94×0.5+0.44")2000 labels/roll (set of 1 rectangle and 1 circle)Log in for availabilityRoll of 1$236.20Supplier Number: 807656016450
- 76273-808Deep freeze labelDeep freeze labelWhite3"23.9×12.7+11.1 mm (0.94×0.5+0.44")4000 labels/roll (set of 1 rectangle and 1 circle)Log in for availabilityRoll of 1$472.66Supplier Number: 807656016467
- 76273-810Deep freeze labelDeep freeze labelWhite1"28.6×19.1+11.1 mm (1.12×0.75+0.44")2000 labels/roll (set of 1 rectangle and 1 circle)Log in for availabilityRoll of 1$243.30Supplier Number: 807656016474
- 76273-812Deep freeze labelDeep freeze labelWhite3"28.6×19.1+11.1 mm (1.12×0.75+0.44")4000 labels/roll (set of 1 rectangle and 1 circle)Log in for availabilityRoll of 1$486.85Supplier Number: 807656016481
- 76273-814Deep freeze labelDeep freeze labelWhite1"30×7.1+9.4 mm (1.18×0.28+0.37")1000 labels/roll (set of 2 rectangles and 1 circle)Log in for availabilityRoll of 1$118.39Supplier Number: 807656016498
- 76273-816Deep freeze labelDeep freeze labelWhite3"30×7.1+9.4 mm (1.18×0.28+0.37")1000 labels/roll (set of 2 rectangles and 1 circle)Log in for availabilityRoll of 1$354.44Supplier Number: 807656016504
Rectangle removable deep freeze labels for thermal transfer printers
Product Information
- ColorCore sizeDimensionsPackagingAvailabilityPrice
- 76273-712Deep freeze labelDeep freeze labelWhite1"6.4×25.4 mm (0.25×1")10000 labels/roll (10 labels across)Log in for availabilityRoll of 1$478.60Supplier Number: 807656015958
- 76273-714Deep freeze labelDeep freeze labelWhite3"6.4×25.4 mm (0.25×1")10000 labels/roll (10 labels across)Log in for availabilityRoll of 1$478.60Supplier Number: 807656015965
- 76273-716Deep freeze labelDeep freeze labelWhite1"22.2×11 mm (0.87×0.44")2000 labels/roll (1 label across)Log in for availabilityRoll of 1$160.50Supplier Number: 807656015972
- 76273-718Deep freeze labelDeep freeze labelWhite3"22.2×11 mm (0.87×0.44")5000 labels/roll (1 label across)Log in for availabilityRoll of 1$397.14Supplier Number: 807656027623
- 76273-720Deep freeze labelDeep freeze labelWhite1"22.2×203.2 mm (0.87×8")250 labels/roll (1 label across)Log in for availabilityRoll of 1$138.11Supplier Number: 807656015989
- 76273-722Deep freeze labelDeep freeze labelWhite1"25.4×7 mm (1×0.28")2000 labels/roll (1 label across)Log in for availabilityRoll of 1$155.97Supplier Number: 807656015996
- 76273-724Deep freeze labelDeep freeze labelWhite3"25.4×7 mm (1×0.28")4000 labels/roll (1 label across)Log in for availabilityRoll of 1$311.87Supplier Number: 807656016009
- 76273-726Deep freeze labelDeep freeze labelWhite1"25.4×12.7 mm (1×0.5")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$85.94Supplier Number: 807656016016
- 76273-728Deep freeze labelDeep freeze labelWhite3"25.4×12.7 mm (1×0.5")3000 labels/roll (1 label across)Log in for availabilityRoll of 1$257.16Supplier Number: 807656016023
- 76273-730Deep freeze labelDeep freeze labelWhite1"25.4×17.5 mm (1×0.69")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$89.60Supplier Number: 807656016030
- 76273-732Deep freeze labelDeep freeze labelWhite3"25.4×17.5 mm (1×0.69")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$89.60Supplier Number: 807656016047
- 76273-734Deep freeze labelDeep freeze labelWhite1"25.4×19.1 mm (1×0.75")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$93.98Supplier Number: 807656016054
- 76273-736Deep freeze labelDeep freeze labelWhite3"25.4×19.1 mm (1×0.75")5000 labels/roll (1 label across)Log in for availabilityRoll of 1$470.54Supplier Number: 807656016061
- 76273-738Deep freeze labelDeep freeze labelWhite1"25.4×25.4 mm (1×1")2000 labels/roll (1 label across)Log in for availabilityRoll of 1$204.41Supplier Number: 807656016078
- 76273-740Deep freeze labelDeep freeze labelWhite3"25.4×25.4 mm (1×1")2000 labels/roll (1 label across)Log in for availabilityRoll of 1$204.41Supplier Number: 807656016085
- 76273-742Deep freeze labelDeep freeze labelWhite1"25.4×44.5 mm (1×1.75")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$147.15Supplier Number: 807656016092
- 76273-744Deep freeze labelDeep freeze labelWhite3"25.4×44.5 mm (1×1.75")3000 labels/roll (1 label across)Log in for availabilityRoll of 1$441.02Supplier Number: 807656016108
- 76273-746Deep freeze labelDeep freeze labelWhite1"25.4×44.5 mm (1×1.75")3000 labels/roll (3 labels across)Log in for availabilityRoll of 1$441.21Supplier Number: 807656016115
- 76273-748Deep freeze labelDeep freeze labelWhite3"25.4×44.5 mm (1×1.75")10000 labels/roll (3 labels across)Log in for availabilityRoll of 1$1,472.40Supplier Number: 807656016122
- 76273-750Deep freeze labelDeep freeze labelWhite1"25.4×50.8 mm (1×2")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$204.41Supplier Number: 807656016139
- 76273-752Deep freeze labelDeep freeze labelWhite3"25.4×50.8 mm (1×2")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$204.41Supplier Number: 807656016146
- 76273-754Deep freeze labelDeep freeze labelWhite1"32.5×15.9 mm (1.28×0.62")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$95.90Supplier Number: 807656016153
- 76273-756Deep freeze labelDeep freeze labelWhite3"32.5×15.9 mm (1.28×0.62")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$95.90Supplier Number: 807656016160
- 76273-758Deep freeze labelDeep freeze labelWhite1"33×15.9 mm (1.3×0.62")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$95.90Supplier Number: 807656016177
- 76273-760Deep freeze labelDeep freeze labelWhite3"33×15.9 mm (1.3×0.62")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$95.90Supplier Number: 807656016184
- 76210-466Deep freeze labelDeep freeze labelWhite1"38.1×19.1 mm (1.5×0.75")1000 labels/roll (1 label across)Log in for availabilityRoll of 1,000$116.09Supplier Number: 807656031132
- 76210-468Deep freeze labelDeep freeze labelWhite3"38.1×19.1 mm (1.5×0.75")2000 labels/roll (1 label across)Log in for availabilityRoll of 2,000$230.11Supplier Number: 807656031149
- 76273-762Deep freeze labelDeep freeze labelWhite1"38.1×63.5 mm (1.5×2.5")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$321.22Supplier Number: 807656016191
- 76273-764Deep freeze labelDeep freeze labelWhite3"38.1×63.5 mm (1.5×2.5")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$321.22Supplier Number: 807656016207
- 76273-766Deep freeze labelDeep freeze labelWhite1"44.5×12.7 mm (1.75×0.5")2000 labels/roll (1 label across)Log in for availabilityRoll of 1$171.53Supplier Number: 807656016214
- 76273-768Deep freeze labelDeep freeze labelWhite3"44.5×12.7 mm (1.75×0.5")6000 labels/roll (1 label across)Log in for availabilityRoll of 1$509.71Supplier Number: 807656016221
- 76273-770Deep freeze labelDeep freeze labelWhite1"44.5×25.4 mm (1.75×1")2000 labels/roll (1 label across)Log in for availabilityRoll of 1$265.56Supplier Number: 807656016238
- 76273-772Deep freeze labelDeep freeze labelWhite3"44.5×25.4 mm (1.75×1")2000 labels/roll (1 label across)Log in for availabilityRoll of 1$265.56Supplier Number: 807656016245
- 76273-798Deep freeze labelDeep freeze labelWhite1"45.7×38.1 mm (1.8×1.5")250 labels/roll (1 label across)Log in for availabilityRoll of 1$152.86Supplier Number: 807656016412
- 76273-800Deep freeze labelDeep freeze labelWhite3"45.7×38.1 mm (1.8×1.5")500 labels/roll (1 label across)Log in for availabilityRoll of 1$305.48Supplier Number: 807656016429
- 76273-646Deep freeze labelDeep freeze labelWhite1"50.8×40.6 mm (2×1.6")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$257.25Supplier Number: 807656016276
- 76273-648Deep freeze labelDeep freeze labelWhite3"50.8×40.6 mm (2×1.6")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$257.25Supplier Number: 807656016283
- 76273-774Deep freeze labelDeep freeze labelWhite1"50.8×25.4 mm (2×1")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$152.41Supplier Number: 807656001562
- 76273-776Deep freeze labelDeep freeze labelOrange1"50.8×25.4 mm (2×1")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$152.41Supplier Number: 807656118154
- 76273-780Deep freeze labelDeep freeze labelOrange3"50.8×25.4 mm (2×1")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$152.41Supplier Number: 807656118161
- 76273-650Deep freeze labelDeep freeze labelWhite1"63.5×25.4 mm (2.5×1")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$190.35Supplier Number: 807656016290
- 76273-652Deep freeze labelDeep freeze labelYellow1"63.5×25.4 mm (2.5×1")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$190.35Supplier Number: 807656016306
- 76273-654Deep freeze labelDeep freeze labelBright green1"63.5×25.4 mm (2.5×1")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$190.35Supplier Number: 807656016313
- 76273-656Deep freeze labelDeep freeze labelRed1"63.5×25.4 mm (2.5×1")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$190.35Supplier Number: 807656016320
- 76273-658Deep freeze labelDeep freeze labelWhite3"63.5×25.4 mm (2.5×1")3000 labels/roll (1 label across)Log in for availabilityRoll of 1$570.96Supplier Number: 807656016337
- 76273-660Deep freeze labelDeep freeze labelYellow3"63.5×25.4 mm (2.5×1")3000 labels/roll (1 label across)Log in for availabilityRoll of 1$570.96Supplier Number: 807656016344
- 76273-782Deep freeze labelDeep freeze labelBright green3"63.5×25.4 mm (2.5×1")3000 labels/roll (1 label across)Log in for availabilityRoll of 1$570.96Supplier Number: 807656016351
- 76273-784Deep freeze labelDeep freeze labelRed3"63.5×25.4 mm (2.5×1")3000 labels/roll (1 label across)Log in for availabilityRoll of 1$570.96Supplier Number: 807656016368
- 76273-786Deep freeze labelDeep freeze labelWhite1"67.1×7 mm (2.64×0.28")2000 labels/roll (1 label across)Log in for availabilityRoll of 1$170.48Supplier Number: 807656016375
- 76273-788Deep freeze labelDeep freeze labelWhite3"67.1×7 mm (2.64×0.28")4000 labels/roll (1 label across)Log in for availabilityRoll of 1$340.79Supplier Number: 807656016382
- 76273-790Deep freeze labelDeep freeze labelWhite1"67.1×7 mm (2.64×0.28")4000 labels/roll (1 label across)Log in for availabilityRoll of 1$330.86Supplier Number: 807656016399
- 76273-792Deep freeze labelDeep freeze labelWhite3"67.1×7 mm (2.64×0.28")4000 labels/roll (1 label across)Log in for availabilityRoll of 1$330.86Supplier Number: 807656016405
- 76273-794Deep freeze labelDeep freeze labelWhite1"76.2×25.4 mm (3×1")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$193.05Supplier Number: 807656001548
- 76273-796Deep freeze labelDeep freeze labelWhite3"76.2×25.4 mm (3×1")1000 labels/roll (1 label across)Log in for availabilityRoll of 1$193.05Supplier Number: 807656001555
- 76273-802Deep freeze labelDeep freeze labelWhite1"101.6×101.6 mm (4×4")300 labels/roll (1 label across)Log in for availabilityRoll of 1$271.50Supplier Number: 807656016436
- 76273-804Deep freeze labelDeep freeze labelWhite3"101.6×101.6 mm (4×4")500 labels/roll (1 label across)Log in for availabilityRoll of 1$452.71Supplier Number: 807656016443