Ionic Strength Adjustor for use with Sodium Ion Selective Electrode, RICCA Chemical Company
RCR4130000-4A
:
- Pack type:Plastic container
- Size:4 L
- Cat. no.:RCR4130000-4A
Ionic Strength Adjustor, for use with Sodium Ion Selective Electrode, packed in a natural poly Container
Colourless liquid.
: Certified suitable for use as an Ionic Strength Adjustor (ISA) for concentration measurement of Sodium Ion (Na+).