WHEATON® Packers, Wide Mouth, Round, HDPE, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Wide mouth HDPE bottles with or without attached caps are available in a range of sizes.
- High density polyethylene construction, natural color
- Good chemical resistance
- Wide mouth design is ideal for use with solids
Delivery: Conveniently supplied with pre-attached white polypropylene screw caps with foamed polyethylene liners.