Order Entry
Canada
ContactUsLinkComponent
WHEATON® Jugs, HDPE, DWK Life Sciences
WHEATON® Jugs, HDPE, DWK Life Sciences
Catalog # 10847-630
undefined
WHEATON® Jugs, HDPE, DWK Life Sciences
Catalog # 10847-630
Supplier Number:  W222341

Specifications

  • Cap Size:
    38-400
  • Capacity:
    3840 ml (128 oz.)
  • Color:
    Natural
  • Cat. no.:
    10847-630
  • Diameter ×H:
    137×295 mm
  • Cap Color:
    White
  • Supplier No.:
    W222341

Specifications

About this item

Manufactured from high density polyethylene.

  • Natural color
  • Good chemical resistance
  • Narrow mouth is ideal for liquids

Conveniently supplied with pre-attached white polypropylene screw cap with polyvinyl liner.