ContactUsLinkComponent
GemTip Cross-Flow II Nebulizer Tip Kit for Optima ICP-OES Instruments, PerkinElmer
GemTip Cross-Flow II Nebulizer Tip Kit for Optima ICP-OES Instruments, PerkinElmer
Catalog # CA97013-636
Supplier:  PerkinElmer
undefined
GemTip Cross-Flow II Nebulizer Tip Kit for Optima ICP-OES Instruments, PerkinElmer
Catalog # CA97013-636
Supplier:  PerkinElmer
Supplier Number:  N0690676
Specifications
  • Description:
    GemTip Cross-Flow II nebulizer tip kit for Optima
  • Cat. no.:
    CA97013-636
  • Supplier No.:
    N0690676
Specifications
About this item
This tip kit is designed for use with GemTip Cross-Flow II Nebulizers in Optima® 2x00/4x00/5x00/7x00 DV ICP-OES series instruments. It includes a clear sapphire sample tip and a red ruby argon tip constructed of PEEK with the gem inserts noted for maximum corrosion resistance.
This tip kit is for use with the GemTip Cross-Flow II Nebulizer for Optima 2x00/4x00/5x00/7x00 (97013-634).