Demountable Quartz Torch, 3-Slot, for Optima ICP-OES Instruments, PerkinElmer
CA97012-866
- Description:3-slot demountable quartz torch for PerkinElmer Optima ICP-OES Instruments
- Cat. no.:CA97012-866
- Supplier No.:N0771500
This 3-Slot quartz torch is used with Optima™ 4300, 5300, and 7300 V ICP-OES series instruments.
This demountable 3-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for use in organic analysis. It is included in the Quick-Change Torch Module for HF Analysis (10136-318), the Organics Sample Introduction Kit (10136-316), and the Quick-Change torch module for oil analysis for Optima 4300/5300/7300 V.