GemCone™ High Dissolved Solids Nebulizer for PerkinElmer Avio and Optima ICP-OES Instruments, PerkinElmer
CA97013-632
- Description:GemCone™ high dissolved solids nebulizer (quick disconnect tubing not included)
- Cat. no.:CA97013-632
- Supplier No.:N0690670
The GemCone™ conespray nebulizer allows you to analyze samples with higher solids concentration than is possible with a Cross-Flow nebulizer.
- Nebulizer is designed for samples with up to 20% high dissolved solids
- Optional quick disconnect tubing increases ease-of-use
- GemCone™ nebulizer is for use with Avio® 200/500 and Optima® 2x00/4x00/5x00/7x00/8x00 ICP-OES series instruments
GemCone™ nebulizers require an end cap for use with Scott Spray Chambers.
This GemCone™ nebulizer is included in the Quick-Change Torch Module for HF Analysis for Optima® 4300V/5300V/7300V (10136-318).