AlphaTec® 23-201 Long-Sleeve PVC Gloves, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
Comfortable, long sleeve PVC glove, with good dexterity and pliability even at low temperatures.
- Rough sandblast finish provides good wet grip
- Medium-duty glove
- Knitted cotton liner, 13 guage
- Food contact approved
The AlphaTec® 23-201 glove features a soft PVC coating which maintains dexterity and pliability, even at low temperatures. It's dipped rough finish makes handling wet and slippery materials easier and safer. It is also heat sealed to offer liquid resistance and dust protection.
Certifications: EN 374; EN 420:2003 + A1:2009; Category lll; REACH compliant.
Packaging: Gloves are packaged 12 pairs per bag, 72 pairs per carton.
Specifications
- Catalog No:
- 75795-486
- 75795-560
- 75795-562
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- Cotton
- Cotton
- Cotton
- Finish:
- Sandblast
- Sandblast
- Sandblast
- Cuff style:
- Extended
- Extended
- Extended