Order Entry
Canada
ContactUsLinkComponent
Reverse Logistics Highway Transport Only Under 49 CFR 173.157 Marking Labels, Labelmaster
Reverse Logistics Highway Transport Only Under 49 CFR 173.157 Marking Labels, Labelmaster
Catalog # 101377-476
Supplier:  Label Master
Reverse Logistics Highway Transport Only Under 49 CFR 173.157 Marking Labels, Labelmaster
Catalog # 101377-476
Supplier:  Label Master
Supplier Number:  L500

Specifications

  • Description:
    Shipping labels
  • Dimensions:
    76.2×127 mm (3×5")
  • Material:
    Semi-gloss paper
  • Thickness:
    5.85 mil.
  • Cat. no.:
    101377-476
  • Legend:
    Reverse Logistics Highway Transport Only Under 49 CFR 173.157 Marking
  • Supplier No.:
    L500

Specifications

About this item

Help ensure compliance when shipping hazardous materials in limited quantities from a retail location back to the manufacturer, supplier, or distribution facility with the Reverse Logistics Highway Transport Markings.

  • Use on retail reverse logistics highway shipments of limited quantities shipped through a private carrier
  • Developed to help comply with 49 CFR 173.157
  • Printed on paper with outstanding chemical and abrasion resistance