ContactUsLinkComponent
DOT Labels 'Explosive Class 1.4 S', Labelmaster
  101376-466
DOT Labels 'Explosive Class 1.4 S', Labelmaster
  101376-466
 :  LEXP14S
  • Description:
    Preprinted dot labels
  • Dimensions:
    100×100 mm (3.94×3.94")
  • Material:
    Semi-gloss paper
  • Thickness:
    5.85 mil.
  • Cat. no.:
    101376-466
  • Packaging:
    On roll
  • Core Size:
    3"
  • Legend:
    Explosive 1.4 S
  • Supplier No.:
    LEXP14S
Help stay in compliance with standards set by the DOT with Explosive Class 1.4 S labels, revised to meet the design requirements of 49 CFR 172.407.
  • Printed with light-fast inks for fade resistance
  • Adhesives formulated for use in various climates and environments
  • Strong holds on corrugated boxes
  • Once affixed, labels won't fade or fall off
  • Outstanding chemcial and abrasion resistance
These labels also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations.