Labels 'This End Up Air', Labelmaster
Catalog # 101376-232
Supplier: Label Master
Specifications
- Description:Preprinted labels
- Dimensions:203.2×101.6 mm (8×4")
- Material:Semi-gloss paper
- Thickness:5.85 mil.
- Cat. no.:101376-232
- Packaging:On roll
- Legend:This End Up
- Supplier No.:IL100R
Specifications
About this item
Get a message across to package handlers with This End Up Air labels.
- Printed on paper
- Outstanding chemical and abrasion resistance
- Strong adhesion to corrugated boxes
Designed to help protect products from mishandling during shipping, these labels include a printed, dotted-line, fold-over feature to mark the top of the box with This End Up. They comply with the requirements in International Air Transport, ICAO, and DOT for marking package orientation on a hazardous materials package.