Labels 'Non-Hazardous Materials', Labelmaster
Catalog # 101377-580
Supplier: Label Master
Specifications
- Description:Preprinted labels
- Dimensions:76.2×76.2 mm (3×3")
- Material:Fluorescent green paper
- Thickness:7.1 mil.
- Cat. no.:101377-580
- Packaging:On roll
- Core Size:3"
- Legend:Non-Hazardous Materials
- Supplier No.:BLT60
Specifications
About this item
Get a message across loud and clear to shipping personnel with Non-Hazardous Materials labels.
- Ultra-bold graphics
- Fluorescent green ink
- Made from heavyweight coated paper
- Contains strong, permanent adhesive
These high-visibility ColorBrite™ warehouse labels command instant attention wherever they’re used.