Order Entry
Canada
ContactUsLinkComponent
Labels 'Defective Do Not Use', Labelmaster
Labels 'Defective Do Not Use', Labelmaster
Catalog # 101376-624
Supplier:  Label Master
Labels 'Defective Do Not Use', Labelmaster
Catalog # 101376-624
Supplier:  Label Master
Supplier Number:  BT338

Specifications

  • Description:
    Preprinted labels
  • Dimensions:
    38.1×50.8 mm (1.5×2")
  • Material:
    Fluorescent red paper
  • Thickness:
    7.1 mil.
  • Environmentally Preferable:
  • Cat. no.:
    101376-624
  • Packaging:
    On roll
  • Legend:
    Defective Do Not Use; By__; Date__
  • Supplier No.:
    BT338

Specifications

About this item

Alert warehouse personnel to important information with Defective Do Not Use labels.

  • Printed on paper
  • Strong adhesion to corrugated boxes

Featuring attention-commanding bright fluorescent red ink, these labels are ideal for inventory control and can be used to expedite shipping, production, and office work. Ample space is provided for personnel to sign off and include the date.