Labels 'Infectious Substance', Labelmaster
Catalog # 101376-354
Supplier: Label Master
Specifications
- Description:Preprinted labels
- Dimensions:100×100 mm (3.94×3.94")
- Material:Semi-gloss paper
- Thickness:5.85 mil.
- Cat. no.:101376-354
- Packaging:On roll
- Legend:Infectious Substance
- Supplier No.:LR17
Specifications
About this item
Ship with confidence when using Worded Infectious Substance labels, revised to meet the design requirements of 49 CFR 172.407.
- Printed with light-fast inks for fade resistance
- Adhesives are formulated for use in various climates and environments
- Once affixed,labels won't fade or fall off
- Outstanding chemical and abrasion resistance
- White and black
In addition to complying with directives set by the DOT, these labels also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations. These Hazard Class 6 labels are made from paper, and are sized at 100×100mm (4×4"). Sold in rolls of 500.