Order Entry
Canada
ContactUsLinkComponent
Radioactive II Labels, Labelmaster
Radioactive II Labels, Labelmaster
Catalog # 101377-914
Supplier:  Label Master
Radioactive II Labels, Labelmaster
Catalog # 101377-914
Supplier:  Label Master
Supplier Number:  HMSL15

Specifications

  • Description:
    Warning labels
  • Dimensions:
    100×100 mm (3.94×3.94")
  • Material:
    PVC-free film
  • Thickness:
    7.1 mil.
  • Cat. no.:
    101377-914
  • Core Size:
    3"
  • Legend:
    Radioactive II
  • Supplier No.:
    HMSL15

Specifications

About this item

Help take the guessing out of shipping label solutions with Worded Radioactive II labels.

  • Revised to meet the design requirements of 49 CFR 172.407
  • Complies with the directives of the Department of Transportation
  • Meet the design and durability standards of 49 CFR, ICAO, and other international air regulations

These Hazard Class 7 labels are printed with light-fast inks for fade resistance, and adhesives are formulated for use in various climates and environments to ensure strong holds on corrugated boxes. Once affixed, they won't fade or fall off. They also feature outstanding chemical and abrasion resistance. White and black, these paper labels are sized at 100×100 mm (4×4"), and sold in rolls of 500.