WHEATON® Replacement Caps for Valumetric™ Graduated Bottles, DWK Life Sciences
Catalog # 10847-612
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Cap Size:38-400
- Environmentally Preferable:
- Cat. no.:10847-612
- Supplier No.:W240541
Specifications
About this item
High-quality replacement caps for Valumetric™ graduated bottles.
- Stress crack resistant
- Superior torque retention
- Excellent sealing characteristics
Manufactured from LDPE. The unique cone design provides a wedge-type seal that not only seals across the top, but also across the inside diameter.