Capillary Sampling Probe for PerkinElmer AA Graphite Furnace Autosamplers, PerkinElmer
Catalog # CA97012-448
Supplier: PerkinElmer
Specifications
- Ø:1.0 mm
- Description:Graphite furnace autosampler probe
- Material:PTFE
- Cat. no.:CA97012-448
- Supplier No.:B0129258
Specifications
About this item
Replacement sampling capillary for Atomic Absorption (AA) instruments equipped with a Graphite Furnace. The PTFE probe is capable of pipetting a variety of corrosive matrices and organic solutions.
- Works well with solvent and acid-resistant sampling components
- PTFE pipette tips and capillaries reduce contamination
- Easy to replace
Compatible with the following, Graphite Furnace Autosamplers: AS-60, AS-70, AS-71, AS-72, AS-800, and AS-900 series.