Palladium 1% (Pd) matrix modifier solution, for graphite furnace AAS
Plastic bottle, 50 ml
Supplier: PerkinElmer
Supplier #: B0190635
Palladium 1% (Pd) matrix modifier solution, for graphite furnace AAS
Catalog # CA97012-036
Supplier: PerkinElmer
CAS Number: 7440-05-3
Jump to:
Product Details & Documents
This 1% Pd as nitrate matrix modifier is used for Atomic Absorption (AA) THGA graphite furnaces.
- CAS Number:7440-05-3
- Formula:Pd
- MDL Number:MFCD00011167
- Molecular Weight:106.420 g/mol
Documents
ghs_B0190635
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Pack type:Plastic bottle
- Size:50 ml



