Order Entry
Canada
ContactUsLinkComponent
 
EMS Swiss Line Standard Tweezer Style 49D, Electron Microscopy Sciences
  76376-072
undefined
EMS Swiss Line Standard Tweezer Style 49D, Electron Microscopy Sciences
  76376-072
 :  7801-49D

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    Style 49D
  • Tip Opening:
    1.5 mm
  • Cat. no.:
    76376-072
  • Overall Length:
    125 mm
  • Tip Width:
    10 mm
  • Tip Length:
    60 mm

 

 

Swiss made standard tweezers have Delrin tips and are temperature resistant up to 115 °C. Delrin is a very soft material that is non scratching, nicking or marring. The tweezers are excellent for use in biotech applications.