Order Entry
Canada
ContactUsLinkComponent
EMS Swiss Line Tech Plus Tweezer Style 7, Electron Microscopy Sciences
EMS Swiss Line Tech Plus Tweezer Style 7, Electron Microscopy Sciences
  76376-044
undefined
EMS Swiss Line Tech Plus Tweezer Style 7, Electron Microscopy Sciences
  76376-044
 :  7800-7

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    Style 7
  • Tip Opening:
    1.5 mm
  • Cat. no.:
    76376-044
  • Overall Length:
    115 mm
  • Tip Width:
    11 mm
  • Tip Length:
    34 mm

 

 

Swiss made tweezers have ultra fine pointed, curved, angled precision tips and edges on the outside rounded off.