Avantor® Partisil Silica, HPLC Columns, 10 µm
: Avantor
Avantor® Partisil® was one of the first commercially available irregular silicas. A large surface area gives it a high loading capacity.
- Irregular porous silica
- 5 and 10 µm particle sizes
- Hichrom high efficiency
- Stainless steel column hardware
Avantor® Partisil® ion-exchange materials are widely referenced and remain one of the most popular choices for analysts.
It is an intermediate carbon load C18 phase used for pharmaceutical, natural products, food, biological and environmental pollutants applications.
- Catalog No:
- 76398-224
- 76398-226
- 76398-240
- 76382-684
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Frequently Bought Together