ContactUsLinkComponent
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Catalog # 76382-600
Supplier:  Avantor
undefined
Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Catalog # 76382-600
Supplier:  Avantor
Supplier Number:  C0001
Specifications
  • Description:
    Column coupler (for use with H0001/H0002)
  • Cat. no.:
    76382-600
Specifications
About this item
Avantor® ACE® PEEK one-piece column couplers are ideal for connecting analytical or semi-preparative columns to stand-alone guard cartridge holders. They can also be used to connect two or more HPLC columns in series or to make similar HPLC connections.
  • Universal connector, ultra-low dead volume
  • Pressure rated to 5000 psi
  • Reusable, female-female connector
  • Inert and biocompatible