ContactUsLinkComponent
LaserShields, 532/810/1064 nm, NoIR Laser Company
LaserShields, 532/810/1064 nm, NoIR Laser Company
Catalog # 76332-456
Supplier:  NOIR MEDICAL
LaserShields, 532/810/1064 nm, NoIR Laser Company
Catalog # 76332-456
Supplier:  NOIR MEDICAL
Supplier Number:  DBD52
Specifications
  • Description:
    532/810/1064 nm lasershields
  • Size:
    Frame 52
  • Cat. no.:
    76332-456
Specifications
About this item
CE and ANSI certified laser protection eyewear.
  • Lightweight polycarbonate
Available in sporty frame #52, or fit-over frame #53.
Certifications: CE Certified LaserShields meet ANSI Z136.1, EN 207/EN 208/EN 60825, and Z87.1 for impact.
Ordering information: Hard-shell case, cleaning cloth, and strap included.