ContactUsLinkComponent
Laser Pointer/Pilot Glare Blue/Green LaserShields, NoIR Laser Company
Laser Pointer/Pilot Glare Blue/Green LaserShields, NoIR Laser Company
Catalog # 76335-178
Supplier:  NOIR MEDICAL
undefined
Laser Pointer/Pilot Glare Blue/Green LaserShields, NoIR Laser Company
Catalog # 76335-178
Supplier:  NOIR MEDICAL
Supplier Number:  PBG60
Specifications
  • Description:
    Laser pointer/pilot glare blue/green lasershields
  • Size:
    Frame 60
  • Cat. no.:
    76335-178
Specifications
About this item
CE and ANSI certified laser protection eyewear.
  • Lightweight polycarbonate
Available in sporty frame #52, fit-over frame #53, or goggle frame #60.
Certifications: CE Certified LaserShields meet ANSI Z136.1, EN 207/EN 208/EN 60825, and Z87.1 for impact.
Ordering information: Hard-shell case, cleaning cloth, and strap included.