WHEATON® FTA® Nucleic Acid Collection Storage Boxes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Storage boxes provide an ideal method for batching/storing FTA® Nucleic acid collection pouches.
- For use with small or large FTA® cards
- Accommodates two rows of 3" FTA® card pouches or one row of 6" FTA® card pouches
- Removable divider for use with smaller pouches
- Water repellent box material allows for longer durability
- For use at RT, 4 °C, –20 °C, or colder
Storage boxes are ideal for nucleic acid collection.
Frequently Bought Together