About this item
These Cubis® II premium precision laboratory balances are ideal for high-performance portfolio of both labs weighing hardware and software.
- Individually configured from different components
- Fully customizable hardware, software, and connectivity
- Customizable modularity, intelligence by design, leading Performance, high speed accuracy, error-free operations, full end to end data integrity, reliable support
- Easy to use manual glass draft shield, safety data transfer
- Removable flat stainless-steel draft shield frame
- Large 7" color touch display including touch, scroll, swipe functionality
- Diverse QApp packages for optional software extension
- Automatic, motorized leveling, gesture control, climate monitoring, built-in ionizer for electrostatic elimination
These balances offer modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
The status center shows all information about your balance and environmental conditions. Automatically execute time-controlled actions for backup.
The Cubis® II balance contains an 'Alibi Memory'. The system automatically stores weighing data in a ring buffer. An electronic signature can be used to sign the final report for weighing processes. Based on a secure password, the combination of username and password is fully compliant with 21CFR.
Different draft sheilds are:
0 - Flat, stainless steel weight pan with no draft shield for weighing modules
A - Automatic, glass motorized draft shield with learning capability for user-friendly operation and easy customization to the changing requirements of different applications
E - Manual glass draft shield for precision balances
I - Identical to the A draft shield, but also includes an integrated ionizer to eliminate interfering electrostatic charges on samples and sample containers
R - Flat, stainless steel weighing pan draft shield (removable, with no glass components) for all precision balances
U - Manual glass analytical draft shield chamber, with smooth-action doors that open wide and provide unimpeded access to the weighing chamber without interfering braces.
Specifications
- Catalog No:
- CA76333-802
- CA76334-240
- CA76334-102
- CA76334-146
- CA76334-256
- CA76334-246
- CA76334-244
- CA76334-266
- CA76334-242
- CA76334-264
- CA76334-268
- CA76333-860
- CA76334-100
- CA76334-254
- CA76334-098
- CA76334-252
- CA76333-926
- CA76334-250
- CA76334-248
- CA76334-270
- CA76376-998
- CA76377-034
- Voltage:
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V