Order Entry
Canada
ContactUsLinkComponent
Distillation Column, Plain, DWK Lifesciences
Distillation Column, Plain, DWK Lifesciences
  KT286000-0022
undefined
Distillation Column, Plain, DWK Lifesciences
  KT286000-0022
 :  286000-0022

 

  • ST Joint:
    14/20
  • Description:
    Distillation column, height 100 mm
  • Cat. no.:
    KT286000-0022
  • Overall Length:
    190 mm
  • Supplier No.:
    286000-0022

 

 

These columns are plain fractionating column with indents to support packing.

  • Variety of packing materials may be used

Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.