KIMBLE® NMR Sealing Manifold, DWK Life Sciences
Catalog #: KT897047-0001
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:NMR sealing manifold
- Cat. no.:KT897047-0001
- Supplier No.:897047-0001
Specifications
About this item
NMR sealing manifold provides an easy way to evacuate and tip-off NMR tubes without additional sealing to other glass apparatus.
- Utilize Kimble® HI-VAC® valves
- All O-rings supplied are FKM
- O-ring size shaft - tip: 110-011
NMR manifolds feature a BEVEL-SEAL™ threaded connection and a specially designed cap with an aluminum shield for heat deflection.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought