KIMBLE® Straight-Bore Stopcock with PTFE Plug, DWK Life Sciences
The more exaggerated taper of this stopcock permits easier use at higher-than-ambient temperatures.
- With a 1:5 taper PTFE plug and a threaded pressure nut
: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Frequently Bought Together