Distillation Adapters, 105° Bend, with Drip Tip, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Bent 105° distillation adapters.
- Standard taper inner drip joint at the bottom and standard taper outer joint at the top
- Joints are the same size
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought