Order Entry
Canada
Orders LinkContactUsLinkComponent
pH Buffer Solutions, NIST/DIN

pH Buffer Solutions, NIST/DIN

pH Buffer Solutions, NIST/DIN
Supplier:  Mettler Toledo
Certificates
pH Buffer Solutions, NIST/DIN
pH Buffer Solutions, NIST/DIN

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

The NIST/DIN pH buffers are specified to three decimal places, for example 9.180, offering the highest possible pH precision.

  • Used for calibration/verification purposes
  • Uncertainty: ±0.015 pH

A detailed test certificate is available for every bottle and guarantees the tested values and traceability.

Single-use sachets, filled under controlled conditions, guarantee a fresh and uncontaminated solution for every calibration. The round and large opening of the sachets allows for easy sensor insertion and makes calibration a quick and simple task.

Certifications: NIST traceable.

Packaging: Packed in either sachets or bottles.

Documents

Product Family Options

Product Information

  • Pack type
    Size
    pH
    Availability
    Price
  • 11278-348
    NIST/DIN buffer solution, shelf life: 12 months
    NIST/DIN buffer solution, shelf life: 12 months
    Bottle
    250 ml
    10.012
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $82.00
    $82.00
    Each (250ml)
       $82.00

    Supplier Number: 51350058
  • 11278-352
    NIST/DIN buffer solution, shelf life: 24 months
    NIST/DIN buffer solution, shelf life: 24 months
    Bottle
    250 ml
    6.865
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $85.70
    $85.70
    Each (250ml)
       $85.70

    Supplier Number: 51350054
  • 11278-354
    NIST/DIN buffer solution, shelf life: 12 months
    NIST/DIN buffer solution, shelf life: 12 months
    Bottle
    250 ml
    9.180
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $85.70
    $85.70
    Each (250ml)
       $85.70

    Supplier Number: 51350056

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.