ContactUsLinkComponent
Reflection and Refraction Tank
Reflection and Refraction Tank
Catalog # 470320-750
undefined
Reflection and Refraction Tank
Catalog # 470320-750
Supplier Number:  RRT002
Specifications
  • Description:
    Reflection refraction demonstrator
  • Cat. no.:
    470320-750
Specifications
About this item
Classical demonstration of refraction and internal reflection.
  • Easy to position laser pointer
  • Includes manual
This product effectively demonstrates the laws of reflection and refraction. The 16 cm diameter acrylic water tank has a 360 degree scale and a moveable arm carrying a laser activated by a push-button. Thumb screws on the side of the laser harness can be adjusted to finely position the laser beam.
Ordering information: Requires two AAA batteries (not included). Includes activity guide.