Led Array
Led array
Catalog # 470320-738
Supplier: United Scientific Supplies
Supplier #: LEDA01
Jump to:
Product Details & Documents
Complete demonstration for colors and LED properties.
- Series of ten single-color LED’s cover the visible spectrum, plus one 'white' wide spectrum LED
- Each LED can be selected individually, or all together
- Demonstrates the color-wavelength relationship
The vertically-aligned set of LEDs make an easy and dramatic demonstration of the relationship between the color and wavelength of light. The spectral range of each LED can be investigated with a spectrometer, or just viewed for comparisons with each other and with the 'white' LED spectrum.
Each LED can be turned on individually with the rotary switch on the top of the column.
Delivery information: Includes power supply.
Documents
Specifications
- Description:Led array