Linearity Urine/Fluids Chemistry, AUDIT Microcontrols
Catalog # 10148-914
Supplier: Audit Microcontrols
CAS Number:
Specifications
- Description:Linearity FD urine/fluids chemistry
- Size:10×3ml
- Cat. no.:10148-914
- Stability:Set 1: 5 Days, Set 2: 5 days when stored at 2...8 °C
- Analyte:Amylase, Calcium, Chloride, Creatinine, Glucose, Magnesium, Microalbumin, Microprotein, Osmolality, Phosphorus, Potassium, Sodium, Urea Nitrogen, Uric Acid
- Type:Freeze dried
- Supplier No.:K723M-5
Specifications
About this item
Urine/Fluids chemistry linearity is assayed quality control material consisting of two sets of five levels of aqueous buffer. The analytes within this kit compose two sets of vials, Set 1 and Set 2. Refer to product package insert.
Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.