- Accuracy:±1.0% rdg 8 digits (DC voltage);
±1.0% rdg 3 digits (DC current);
±1.5% rdg 10 digits (AC voltage);
±3.0% rdg 5 digits (AC current);
±1.5% ± 5 digits (frequency) - Frequency:9.999 Hz - 9.999 MHz
- Temperature range:‒45...750 °C (‒50...1382 °F)
- Description:Digi-Sense™ Heavy-duty industrial digital multimeter with Bluetooth® connectivity
- Voltage AC:0.4 - 1000 V
- Current AC:0.0004 - 10 A
- Capacitance accuracy:±3.0% rdg ± 5 digits
- Category rating:CAT IV-600 V, CAT III-1000 V
- Voltage DC:0.4 - 1000 V
- Current DC:0.0004 - 10 A
- Temperature accuracy:±3.0% rdg ± 5 °C (9 °F)
- Capacitance:4000000 - 40 nF
- Resistance accuracy:±1.2% rdg ± 4 digits
- Display:Backlit LCD
- Resistance:400Ω - 40 MΩ
- Cat. no.:76360-052
- L×W×H:2.25×3.25×7"
View, store, and share real-time data from a safe distance on your smartphone.
- Get the most out of your data with our user-friendly mobile app
- Unlimited data storage based on smart device capacity
- Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards
- Rugged double-molded housing provides increased durability
- Bright, large backlit 4000-count LCD permits viewing in low-light conditions
- Tilt stand offers easy hands-free viewing
- IP67 waterproof rated
- CAT IV-600 V, CAT III-1000 V safety rating
- Continuity
- Diode Tester
- Duty Cycle
- True Rms
Using the free Digi-Sense™ D/S Meter app, this heavy-duty industrial digital multimeter with Bluetooth® technology allows you to transmit data directly to your Android or iOS device. Now you can view data at a safe distance from dangerous equipment or hazardous locations and even use your smart device as a real-time secondary display for checking measurements, eliminating the need to write down readings on paper. The results are automatically stored in the app with a date-and-time stamp and can be saved as a CSV file and emailed for future reference, manipulation, or analysis to help determine trends and conditions over a long period of time. Use the app to attach photos and notes to the records creating a clear reference point for your work - ideal for record keeping at large sites.
The handheld digital multimeter measures AC/DC voltage, AC/DC current, resistance, frequency, capacitance, and continuity. Special functions include auto ranging, data Hold, Max/Min readings, diode test, audible continuity check, relative mode, duty cycle, low-battery indication, and auto power-off. The meter also offers True RMS for more accurate current readings, temperature measurement with a type K thermocouple, and analog bar graph for viewing trends.