Metal Solvent Resistant Pipettes, Labsciences
Supplier: LAB SCIENCE CA
Certificates
About this item
These metal fixed volume pipettes are manufactured for pipetting fuel, oil and solvents.
- Fully autoclavable to avoid cross-contamination
- Rugged metal body that does not break or easily loose its calibration
- Lightweight metal construction with soft plunger movement
- Precision machined for high accuracy and repeatibility
The compact size makes the pipettes ideal for field work.
Certifications: Calibrated in accordance with DIN 12650 Standards.
Customers Who Bought This Also Bought