Order Entry
Canada
ContactUsLinkComponent
Linearity Beta-Hydroxybutyric Acid, AUDIT Microcontrols
Linearity Beta-Hydroxybutyric Acid, AUDIT Microcontrols
Catalog # 10148-922
CAS Number:  
Linearity Beta-Hydroxybutyric Acid, AUDIT Microcontrols
Catalog # 10148-922
Supplier Number:  K728M-5
CAS Number:  

Specifications

  • Description:
    Linearity LQ Beta-Hydroxybutyric Acid
  • Size:
    5×1 ml
  • Cat. no.:
    10148-922
  • Stability:
    7 days when stored at 2...8 °C
  • Analyte:
    Beta-Hydroxybutyric Acid
  • Type:
    Liquid
  • Supplier No.:
    K728M-5

Specifications

About this item

Beta-Hydroxybutyric acid Linearity is assayed quality control material consisting of five levels of human based serum.

Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.