Vacuum Trap, 35/20 Ball Joints, Chemscience
Catalog # CA10203-026
Supplier: CHEM SCIENCE INC CA
Specifications
- ST Joint No.:45/50
- Description:Vacuum trap, 35/20 ball joints
- Cat. no.:CA10203-026
- Supplier No.:SI-V0415045
Specifications
About this item
For use with vacuum manifolds having 35/20 socket joints.
Ordering information: Please order the pump adapter and socket clamp separately.