Stopcocks, 1:5 PTFE, T-Bore, Chemscience
Stopcock with three stems sealed 90° apart.
PTFE plug has a "T" shape bore to connect any two or all three stems.
Stopcock with three stems sealed 90° apart.
PTFE plug has a "T" shape bore to connect any two or all three stems.
Product Family Options
- DescriptionStem O.D.Bore sizePlug size
- : CS-S0161530
- : CS-S0161635