Orders LinkContactUsLinkComponent
Stopcocks, PTFE Straight Bore, Chemscience
Stopcocks, PTFE Straight Bore, Chemscience
Supplier:   CHEM SCIENCE INC CA
Stopcocks, PTFE Straight Bore, Chemscience
Certificates
Stopcocks, PTFE Straight Bore, Chemscience
Stopcocks, PTFE Straight Bore, Chemscience
3 Options Available Below
Product Details & Documents
With 1:5 PTFE plug and a threaded pressure nut.
Stopcock barrels are polished to prevent PTFE plug from scratch.
Product Family Options
Product Information
  • Bore size
    Plug size
    Availability
    Price
  • CA10202-960
    Stopcock, PTFE straight bore
    Stopcock, PTFE straight bore
    2 mm
    11/25
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $84.22
    $84.22
    Case of 1
       $84.22
    Supplier Number: CS-S0101502
  • CA10202-962
    Stopcock, PTFE straight bore
    Stopcock, PTFE straight bore
    4 mm
    15.2/30
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $127.48
    $127.48
    Case of 1
       $127.48
    Supplier Number: CS-S0101504
  • CA10202-964
    Stopcock, PTFE straight bore
    Stopcock, PTFE straight bore
    6 mm
    16/35
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $158.03
    $158.03
    Case of 1
       $158.03
    Supplier Number: CS-S0101506