Order Entry
Canada
Orders LinkContactUsLinkComponent
Difco™ MIL Medium, BD

Difco™ MIL Medium, BD

MIL medium, 500 g
Catalog # CA90000-962
Supplier:   BD
Supplier #:  218041
Difco™ MIL Medium, BD
Catalog # CA90000-962
Supplier:  BD
CAS Number:  
Difco™ MIL Medium, BD

MIL medium, 500 g

CAS Number:  
Price
$474.00
Log in for availability
Each (500g)
Product Details & Documents

Motility indole lysine medium is used for differentiating Enterobacteriaceae based on motility, lysine decarboxylation, lysine deamination and indole production.

  • Dehydrated appearance: light beige, free-flowing, homogeneous
  • Prepared appearance: reddish purple, clear, semisolid
  • pH: 6.6 ±0.2 (at 25 °C)
Specifications

Specifications

  • Description:
    MIL medium
  • Size:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.