Solvent Still Heads, With PTFE Stopcock, Chemscience, SATI International Scientific
Supplier: CHEM SCIENCE INC CA
Certificates
About this item
Syringe port has 14/20 outer joint for a rubber septum and a 4 mm PTFE stopcock.
Solvent is drained through 2 mm T-bore PTFE stopcock. The drainage arm has a 14/20 inner joint.
Customers Who Bought This Also Bought