Solvent Still Heads, Chemscience, SATI International Scientific
Supplier: CHEM SCIENCE INC CA
Certificates
About this item
These solvents include a drip tip on the delivery port other than 14/20 inner joint.
With 4 mm straight bore stopcock in the upper portion for easy syringe needle access.
Customers Who Bought This Also Bought