Order Entry
Canada
ContactUsLinkComponent
BD BBL™ Todd Hewitt Broth with Gentamicin and Nalidixic Acid, Prepared Media, BD Diagnostics
BD BBL™ Todd Hewitt Broth with Gentamicin and Nalidixic Acid, Prepared Media, BD Diagnostics
Catalog # CA90005-220
Supplier:  BD
CAS Number:  
BD BBL™ Todd Hewitt Broth with Gentamicin and Nalidixic Acid, Prepared Media, BD Diagnostics
Catalog # CA90005-220
Supplier:  BD
Supplier Number:  299486
CAS Number:  

Specifications

  • Description:
    Todd Hewitt broth with Gentamicin and Nalidixic Acid Tube
  • Size:
    Carton of 100 size K tubes
  • Cat. no.:
    CA90005-220
  • Shelf Life:
    365
  • Supplier No.:
    299486

Specifications

About this item

Todd Hewitt broth with gentamicin and nalidixic acid is used for the selective enrichment of group B streptococci (Streptococcus agalactiae).

  • Do not open until ready to use
  • Medium is highly nutritious due to its content of peptones, dextrose and salts
  • Store tubes in the dark at 2 to 8 °C

Selective enrichment broths include the advantages of both enrichment and selection by providing conditions conducive to the growth of group B streptococci while inhibiting the growth of contaminants.