KIMBLE® Separatory Funnels, Squibb Type, DWK Life Sciences
About this item
KONTES® Squibb separatory funnels with PTFE stopcock and standard taper joint.
- Plain stem, but may be used with PTFE BEVEL-SEAL™ inlet adapter (Note stem diameter for a proper fit when using this adapter)
- With an 1:5 taper PTFE stopcock plug
- Manufactured from 33 expansion, low extractable borosilicate glass
KT636031-1000 is supplied without a stopper. KIMBLE® separatory funnels, squibb type (KT636030 series) is supplied with a glass stopper.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
8 Options Available Below
Product Details & Documents
KONTES® Squibb separatory funnels with PTFE stopcock and standard taper joint.
- Plain stem, but may be used with PTFE BEVEL-SEAL™ inlet adapter (Note stem diameter for a proper fit when using this adapter)
- With an 1:5 taper PTFE stopcock plug
- Manufactured from 33 expansion, low extractable borosilicate glass
KT636031-1000 is supplied without a stopper. KIMBLE® separatory funnels, squibb type (KT636030 series) is supplied with a glass stopper.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- CapacityHeightAvailabilityPrice
- KT636030-0060Seporatory funnel with 16 stopperSupplier Number: 636030-0060
- KT636030-0125Seporatory funnelSupplier Number: 636030-0125
- KT636030-0250Seporatory funnel with 24/25 stopperSupplier Number: 636030-0250
- KT636030-0500Seporatory funnelSupplier Number: 636030-0500
- KT636030-1000Seporatory funnelSupplier Number: 636030-1000
- KT636031-1000Seporatory funnel without stopperSupplier Number: 636031-1000
- KT636030-2000Seporatory funnelSupplier Number: 636030-2000
- KT636030-4000Seporatory funnelSupplier Number: 636030-4000