KIMBLE® Variable Reflux Distillation Head with PTFE Plugs, Small, DWK Life Sciences
420 mm, Variable reflux distillation head with PTFE plugs (small), 24/40
Catalog # KT518530-2440
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 518530-2440
KIMBLE® Variable Reflux Distillation Head with PTFE Plugs, Small, DWK Life Sciences
Catalog # KT518530-2440
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
Rotation of cold finger condenser allows collection of distillate or return to flask.
- Stopcocks allow removal of the product or a receiver change without interrupting the distillation
- Lower receiver joint has a drip tip
- Standard Taper 10/30 outer side joint accepts a 75 mm immersion thermometer
- Stopcock plugs are PTFE
- Manufactured from 33 expansion, low extractable borosilicate glass
Top and bottom plugs are KT821001-0004 and the side is KT824001-0004.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Specifications
- Description:Variable reflux distillation head with PTFE plugs (small)
- Height:420 mm
- [ST] joint size:24/40
Accessories
Product Information
- Catalog #AvailabilityPrice
- Cold finger joint with lower drip tip, 305 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



