Difco™ Buffered Listeria Enrichment Broth Base, BD Diagnostics
Catalog # CA90004-134
Supplier: BD
CAS Number:
Specifications
- Description:Difco™ buffered Listeria enrichment broth base
- Size:500 g
- Cat. no.:CA90004-134
- Supplier No.:290720
Specifications
About this item
Buffered Listeria enrichment broth base used for the isolation of Listeria species.