Anti-IgG Sheep Polyclonal Antibody
Catalog # 76334-052
Supplier: Biosensis
Supplier #: S-1759-100
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Sheep serum was obtained from non-immunized animals. This whole serum can be used as negative control antibody for various immunological techniques where sheep primary antibodies (whole serum) are used.
Applications: Use as negative control for sheep primary antibodies (whole serum) at matching antibody dilution.
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Specifications
- Cat. No.:76334-052
- Clonality:Polyclonal
- Host:Sheep
- Antigen symbol:IgG
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



