BBL™ Phenol Red Broth, BD Life Sciences
Supplier: BD
Certificates
About this item
Phenol red broth with carbohydrates are used for the determination of fermentation reactions in the differentiation of microorganisms.
- Complete medium
- Detect acid production
- Used as negative control for fermentation
Most of the end products of carbohydrate fermentation are organic acids which, in the presence of phenol red, produce a color change in the medium from red to yellow.
If gas is produced during the fermentation reaction, it is collected in the inverted Durham tube.
Frequently Bought Together