Specifications
- Description:BBL™ cooked meat medium
- Size:100 g
- Cat. no.:CA-90001-374
- Supplier No.:221508
Specifications
About this item
BBL™ cooked meat medium is used for the cultivation of anaerobes in particular clostridia pathogens.
- Supports the growth of most of sporulated, non-spore-bound anaerobes
- Muscle protein present in the granules of cardiac tissue is a source of amino acids and others nutrients
- Growth is indicated by turbidity and presence of gas bubbles
Robertson developed a ground with cooked meat to use in the cultivation of some anaerobes isolated from wounds. The current formulation of the ground with cooked meat is a modification of the Robertson's original formula.
Can be used for the cultivation and conservation of clostridia and for the determination of the proteolytic activity of anaerobes.
This medium provides a favorable environment for the growth of anaerobes. The sulfhydryl groups, which exert the reducing effect, are more available in denatured protein; therefore, the meat particles are cooked for use in the medium.